Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2'-Hydroxy-4',5'-dimethoxyacetophenone 98.0+%, TCI America™
SDP

Catalog No. H12065G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H12065G Supplier TCI America Supplier No. H12065G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 20628-06-2
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.202
MDL Number MFCD00017241
InChI Key KEQHBVWVKYHDCS-UHFFFAOYSA-N
PubChem CID 706870
IUPAC Name 1-(2-hydroxy-4,5-dimethoxyphenyl)ethanone
SMILES CC(=O)C1=CC(=C(C=C1O)OC)OC

Specifications

Melting Point 113°C
Quantity 5 g
Formula Weight 196.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Hydroxy-4′,5′-dimethoxyacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.