Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxy-4-(methacryloyloxy)benzophenone, 99%

Catalog No. p-7026447
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2244506 5 g
AAB2244514 25 g
AAB2244522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2244506 Supplier Thermo Scientific Chemicals Supplier No. B2244506
Add to Cart

CAS: 2035-72-5 | C17H14O4 | 282.295 g/mol

2-Hydroxy-4-(methacryloyloxy)benzophenone is used in ophthalmic devices, films, coatings, and fibers, cosmetic preparations and toners.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxy-4-(methacryloyloxy)benzophenone is used in ophthalmic devices, films, coatings, and fibers, cosmetic preparations and toners.

Solubility
Insoluble in water.

Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2035-72-5
Molecular Formula C17H14O4
Molecular Weight (g/mol) 282.295
MDL Number MFCD00080561
InChI Key IMNBHNRXUAJVQE-UHFFFAOYSA-N
Synonym 4-benzoyl-3-hydroxyphenyl methacrylate, 4-methacryloxy-2-hydroxybenzophenone, 2-hydroxy-4-methacryloyloxy benzophenone, methacrylic acid 4-benzoyl-3-hydroxyphenyl ester, 3-hydroxy-4-phenylcarbonyl phenyl 2-methylprop-2-enoate, 2-propenoic acid, 2-methyl-, 4-benzoyl-3-hydroxyphenyl ester, acmc-20amfp, 2-hydroxy-4-methacryloxybenzophenone, 2-hydroxy-4-methacryloyloxybenzophenone
PubChem CID 74856
IUPAC Name (4-benzoyl-3-hydroxyphenyl) 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OC1=CC(=C(C=C1)C(=O)C2=CC=CC=C2)O

Specifications

Melting Point 77°C to 79°C
Density 1.18
Quantity 5 g
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 282.3
Percent Purity 99%
Chemical Name or Material 2-Hydroxy-4-(methacryloyloxy)benzophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 218-000-6
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.