Learn More
Medchemexpress LLC 2-Hydroxy-4-methoxybenzaldehyde | 673-22-3 | 152.15 | 10 G
SDP
Catalog No. 5000389687
Encompass_Preferred
2-hydroxy-4-methoxybenzaldehyde is a potent tyrosinase inhibitor. It is an isomer of Vanillin and could be used to synthesize Urolithin M7.
  • Ic50 & target: Tyrosinase
  • In vitro: Inhibits the oxidation of L-3,4-dihydroxyphenylalanine (L-DOPA) by mushroom tyrosinase with an ID50 of 4.3 μg/mL (0.03 mM).
Catalog No. 50-003-89687 Supplier Medchemexpress LLC Supplier No. HYN044510G
Add to Cart
Add to Cart
2-hydroxy-4-methoxybenzaldehyde is a potent tyrosinase inhibitor. It is an isomer of Vanillin and could be used to synthesize Urolithin M7.
  • Ic50 & target: Tyrosinase
  • In vitro: Inhibits the oxidation of L-3,4-dihydroxyphenylalanine (L-DOPA) by mushroom tyrosinase with an ID50 of 4.3 μg/mL (0.03 mM).