Learn More
Medchemexpress LLC 2-Hydroxy-4-methoxybenzaldehyde | 673-22-3 | 152.15 | 1000 G
SDP
Catalog No. 5000389896
Encompass_Preferred
2-Hydroxy-4-methoxybenzaldehyde is identified as a potent tyrosinase inhibitor. It is also an isomer of Vanillin and can be utilized in the synthesis of Urolithin M7. The product is intended for research use only and is not sold to patients.
  • Potent tyrosinase inhibitor.
  • Isomer of Vanillin.
  • Can be used to synthesize Urolithin M7.
  • High purity: 99.96%.
  • Available in various sizes.
  • Provided with data sheet, COA, SDS, and handling instructions.
Catalog No. 50-003-89896 Supplier Medchemexpress LLC Supplier No. HYN04451000G
2-Hydroxy-4-methoxybenzaldehyde is identified as a potent tyrosinase inhibitor. It is also an isomer of Vanillin and can be utilized in the synthesis of Urolithin M7. The product is intended for research use only and is not sold to patients.
  • Potent tyrosinase inhibitor.
  • Isomer of Vanillin.
  • Can be used to synthesize Urolithin M7.
  • High purity: 99.96%.
  • Available in various sizes.
  • Provided with data sheet, COA, SDS, and handling instructions.