Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxy-4-methoxybenzaldehyde, 98%

Catalog No. AAA1297114
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1297114 25 g
AAA1297103 1 g
AAA1297106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1297114 Supplier Thermo Scientific Chemicals Supplier No. A1297114
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 673-22-3 | C8H8O3 | 152.15 g/mol

It is employed in the synthesis of Schiff base ligand. It is applied as a reactant in the synthesis of LPA1R antagonists used in the inhibition of LPA-induced proliferation and contraction of normal human lung fibroblasts. Also used in the synthesis of tyrosine kinase 6 proteinase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the synthesis of Schiff base ligand. It is applied as a reactant in the synthesis of LPA1R antagonists used in the inhibition of LPA-induced proliferation and contraction of normal human lung fibroblasts. Also used in the synthesis of tyrosine kinase 6 proteinase inhibitors.

Solubility
Solubility in methanol is almost transparent. Insoluble in water.

Notes
Air sensitive. Store away from oxidizing agents, bases and air/oxygen. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 673-22-3
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00003327
InChI Key WZUODJNEIXSNEU-UHFFFAOYSA-N
Synonym 4-methoxysalicylaldehyde, benzaldehyde, 2-hydroxy-4-methoxy, 4-methoxysalicyaldehyde, 2-hydroxy-4-methoxy-benzaldehyde, o-hydroxy-p-methoxybenzaldehyde, p-anisaldehyde, 2-hydroxy, salicylaldehyde, 4-methoxy, 2-hydroxy-p-anisaldehyde, 2-formyl-5-methoxyphenol, unii-2n395p88lw
PubChem CID 69600
IUPAC Name 2-hydroxy-4-methoxybenzaldehyde
SMILES COC1=CC=C(C=O)C(O)=C1

Specifications

Melting Point 40°C to 44°C
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 1072443
Sensitivity Air Sensitive
Formula Weight 152.15
Percent Purity 98%
Chemical Name or Material 2-Hydroxy-4-methoxybenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-604-0
RTECSNumber BZ2810000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.