Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-hydroxy-4-methoxybenzamide

Catalog No. CDS013828 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS013828 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS013828 Supplier Sigma Aldrich Supplier No. CDS013828250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Triisopropyl orthoformate (tipo) is formed during the reaction of dichlorofluoromethane with potassium isopropoxide in isopropyl alcohol
  • Separation of tipo from n-hexane by dense carbon nanotube (cnt) membranes by pervaporation has been reported.triisopropyl orthoformate may be used for the preparation of 2-(diisopropoxymethyl)oxirane

Specifications

CAS 6745-77-3
Molecular Formula C8H9NO3
Linear Formula C8H9NO3
Quantity 100 mL
MDL Number MFCD03840504
Molecular Weight (g/mol) 167.16
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.