Learn More
Medchemexpress LLC 2-Hydroxy-4-methoxybenzoic acid | 2237-36-7 | 99.6% | C8H8O4 | 5 G
SDP
Catalog No. 5000433596
Encompass_Preferred
2-Hydroxy-4-methoxybenzoic acid, or 4-Methoxysalicylic Acid, is a phenolic compound from plants like Hemidesmus indicus, known for its diverse biological activities.
  • Isolated from natural plant sources
  • Exhibits anti-diabetic, hypolipidemic, and hepatoprotective activities
  • Demonstrates anti-snake venom properties
  • Used in research for diabetes, liver, immune-related diseases, and snake venom poisoning
  • Evaluated for cytotoxicity against RAW264.7 cells
Catalog No. 50-004-33596 Supplier Medchemexpress LLC Supplier No. HY756255G
Add to Cart
Add to Cart
2-Hydroxy-4-methoxybenzoic acid, or 4-Methoxysalicylic Acid, is a phenolic compound from plants like Hemidesmus indicus, known for its diverse biological activities.
  • Isolated from natural plant sources
  • Exhibits anti-diabetic, hypolipidemic, and hepatoprotective activities
  • Demonstrates anti-snake venom properties
  • Used in research for diabetes, liver, immune-related diseases, and snake venom poisoning
  • Evaluated for cytotoxicity against RAW264.7 cells