Learn More
Medchemexpress LLC 2-Hydroxy-4-methoxybenzoic acid | 2237-36-7 | MFCD00002450 | 99.6% | C8H8O4 | 100 G
SDP
Catalog No. 5000433868
Encompass_Preferred
2-Hydroxy-4-methoxybenzoic acid is a phenolic compound isolated from *Hemidesmus indicus* and other plants. It exhibits anti-diabetic, hypolipidemic, hepatoprotective, and anti-snake venom activities. This compound can be used in studies related to diabetes and its complications, liver diseases, immune-related diseases, and snake venom poisoning.
  • Isolated from Hemidesmus indicus and other plants
  • Exhibits anti-diabetic, hypolipidemic, hepatoprotective, and anti-snake venom activities
  • Useful in studies of diabetes, liver diseases, immune-related diseases, and snake venom poisoning
  • Ships at room temperature in continental US
  • Store at 4°C under nitrogen
  • Store in solvent at -80°C for 6 months, or -20°C for 1 month under nitrogen
Catalog No. 50-004-33868 Supplier Medchemexpress LLC Supplier No. HY75625100G
2-Hydroxy-4-methoxybenzoic acid is a phenolic compound isolated from *Hemidesmus indicus* and other plants. It exhibits anti-diabetic, hypolipidemic, hepatoprotective, and anti-snake venom activities. This compound can be used in studies related to diabetes and its complications, liver diseases, immune-related diseases, and snake venom poisoning.
  • Isolated from Hemidesmus indicus and other plants
  • Exhibits anti-diabetic, hypolipidemic, hepatoprotective, and anti-snake venom activities
  • Useful in studies of diabetes, liver diseases, immune-related diseases, and snake venom poisoning
  • Ships at room temperature in continental US
  • Store at 4°C under nitrogen
  • Store in solvent at -80°C for 6 months, or -20°C for 1 month under nitrogen