Learn More
Medchemexpress LLC 2-Hydroxy-4-methoxybenzoic acid (4-methoxysalicylic acid) | 2237-36-7 | 99.6% | C8H8O4 | 10 G
SDP
Catalog No. 5000434347
Encompass_Preferred
2-Hydroxy-4-methoxybenzoic acid (4-Methoxysalicylic Acid) is a phenolic compound isolated from Hemidesmus indicus and other plants. It exhibits anti-diabetic, hypolipidemic, hepatoprotective, and anti-snake venom activities.
  • Used in the study of diabetes and its complications
  • Used in the study of liver diseases
  • Used in the study of immune-related diseases
  • Used in the study of snake venom poisoning
Catalog No. 50-004-34347 Supplier Medchemexpress LLC Supplier No. HY7562510G
2-Hydroxy-4-methoxybenzoic acid (4-Methoxysalicylic Acid) is a phenolic compound isolated from Hemidesmus indicus and other plants. It exhibits anti-diabetic, hypolipidemic, hepatoprotective, and anti-snake venom activities.
  • Used in the study of diabetes and its complications
  • Used in the study of liver diseases
  • Used in the study of immune-related diseases
  • Used in the study of snake venom poisoning