Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2'-Hydroxy-4'-methoxychalcone

Catalog No. IDF000571G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IDF000571G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. IDF000571G Supplier Sigma Aldrich Supplier No. IDF000571G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-bromo-n-methylaniline can be synthesized via the reduction of 2-bromoformylanilide.2-bromo-n-methylaniline can be used to synthesize benzimidazole derivatives, via one pot n-arylation of amides/cyclic amides in the presence of copper iodide nanoparticles
  • It can also be used in the synthesis of alkyltelluro-substituted aromatic amines, which can show radical trapping ability

Specifications

CAS 39273-61-5
Molecular Formula C16H14O3
Linear Formula C16H14O3
MDL Number MFCD00017173
Quantity 1 g
Molecular Weight (g/mol) 254.28
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.