Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxy-4-methyl-5-nitropyridine, 97%, Thermo Scientific™

Catalog No. AAL1986703
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1986703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1986703 Supplier Thermo Scientific Chemicals Supplier No. L1986703

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21901-41-7
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.125
MDL Number MFCD00010690
InChI Key AIEHUZHKFUNHCJ-UHFFFAOYSA-N
Synonym 2-hydroxy-4-methyl-5-nitropyridine, 4-methyl-5-nitropyridin-2-ol, 2-hydroxy-5-nitro-4-picoline, 2-hydroxy-5-nitro-4-methyl pyridine, 2-hydroxy-5-nitro-4-methylpyridine, 4-methyl-5-nitropyridin-2 1h-one, 4-methyl-5-nitro-2-pyridinol, 4-methyl-5-nitro-1,2-dihydropyridin-2-one, 4-methyl-5-nitro-pyridin-2-ol, 2 1h-pyridinone, 4-methyl-5-nitro
PubChem CID 345371
IUPAC Name 4-methyl-5-nitro-1H-pyridin-2-one
SMILES CC1=CC(=O)NC=C1[N+](=O)[O-]

Specifications

Melting Point 188°C to 190°C
Odor Odorless
Quantity 1 g
Beilstein 136900
Formula Weight 154.13
Percent Purity 97%
Chemical Name or Material 2-Hydroxy-4-methyl-5-nitropyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.