Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Hydroxy-4-methylth 100g | 583-91-5 | 100 G
SDP

Catalog No. 5000228882
Encompass_Preferred

2-Hydroxy-4-(methylthio)butyric acid is an alpha-hydroxy derivative of methionine supplied for laboratory research use. It is commonly used in biochemical studies and animal nutrition research and is provided with supporting documentation for safe handling and quality verification.

  • For research use only; not for human consumption or clinical use.
  • Packaged as a 100 g package; other sizes are available.
  • Purity ≥88.0% as supplied.
  • Molecular formula C5H10O3S; molecular weight 150.20 g/mol.
  • Safety data sheet and certificate of analysis available.

Catalog No. 50-002-28882 Supplier Medchemexpress LLC Supplier No. HY116688100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Hydroxy-4-(methylthio)butyric acid is an alpha-hydroxy derivative of methionine supplied for laboratory research use. It is commonly used in biochemical studies and animal nutrition research and is provided with supporting documentation for safe handling and quality verification.

  • For research use only; not for human consumption or clinical use.
  • Packaged as a 100 g package; other sizes are available.
  • Purity ≥88.0% as supplied.
  • Molecular formula C5H10O3S; molecular weight 150.20 g/mol.
  • Safety data sheet and certificate of analysis available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.