Learn More
Medchemexpress LLC 2-Hydroxy-4-methylth 10g | 583-91-5 | 150.20 g/mol | C5H10O3S | 10 G
SDP
Catalog No. 5000308179
Encompass_Preferred
2-Hydroxy-4-(methylthio)butyric acid is an alpha-hydroxy analogue of methionine used as an orally active methionine source and as a research reagent in studies of amino acid metabolism and nutritional supplementation. It is supplied as a liquid and is used in formulation and biological assays where methionine precursors are required.
  • Acts as an orally active source of methionine for nutritional studies.
  • Useful in biochemical research on amino acid metabolism and feed supplementation.
  • Supplied as a liquid, facilitating formulation and dosing experiments.
  • Typical purity around ≥88%, suitable for research applications.
  • Available in small research quantities for laboratory use.
Catalog No. 50-003-08179 Supplier Medchemexpress LLC Supplier No. HY11668810G
2-Hydroxy-4-(methylthio)butyric acid is an alpha-hydroxy analogue of methionine used as an orally active methionine source and as a research reagent in studies of amino acid metabolism and nutritional supplementation. It is supplied as a liquid and is used in formulation and biological assays where methionine precursors are required.
  • Acts as an orally active source of methionine for nutritional studies.
  • Useful in biochemical research on amino acid metabolism and feed supplementation.
  • Supplied as a liquid, facilitating formulation and dosing experiments.
  • Typical purity around ≥88%, suitable for research applications.
  • Available in small research quantities for laboratory use.