Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Hydroxy-4-methylth 10mM 1mL | 583-91-5 | 150.20 | C5H10O3S | 1 ML
SDP

Catalog No. 5000307993
Encompass_Preferred

2-Hydroxy-4-(methylthio)butyric acid is an organic acid used as an orally active source of methionine for research applications. Supplied as a ready-to-use 10 mM solution in DMSO (1 mL) and in larger liquid quantities, it is used in studies of methylation, nutrient supplementation, and lactation.

  • Ready-to-use 10 mM solution in DMSO (1 mL) for convenient dosing.
  • Available in bulk liquid quantities for formulation and scale-up.
  • Modulates expression of methylation-related mRNAs such as BHMT, MTR, MAT1A, SAHH, and PCK1.
  • Serves as an orally active methionine precursor for nutritional and physiological studies.
  • Identified by CAS 583-91-5; molecular weight 150.20; formula C5H10O3S.

Catalog No. 50-003-07993 Supplier Medchemexpress LLC Supplier No. HY11668810MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Hydroxy-4-(methylthio)butyric acid is an organic acid used as an orally active source of methionine for research applications. Supplied as a ready-to-use 10 mM solution in DMSO (1 mL) and in larger liquid quantities, it is used in studies of methylation, nutrient supplementation, and lactation.

  • Ready-to-use 10 mM solution in DMSO (1 mL) for convenient dosing.
  • Available in bulk liquid quantities for formulation and scale-up.
  • Modulates expression of methylation-related mRNAs such as BHMT, MTR, MAT1A, SAHH, and PCK1.
  • Serves as an orally active methionine precursor for nutritional and physiological studies.
  • Identified by CAS 583-91-5; molecular weight 150.20; formula C5H10O3S.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.