Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Hydroxy-4-methylth 25g | 583-91-5 | 150.20 g/mol | C5H10O3S | 25 G
SDP

Catalog No. 5000226194
Encompass_Preferred

2-Hydroxy-4-(methylthio)butyric acid (CAS 583-91-5) is the hydroxy analogue of methionine used as a biochemical reagent and reference standard for laboratory research. It is supplied as a light yellow to brown liquid and is intended for research applications under appropriate storage conditions.

  • hydroxy analogue of methionine (CAS 583-91-5)
  • molecular formula C5H10O3S and molecular weight 150.20 g/mol
  • appearance: light yellow to brown liquid
  • purity ≥88.0%
  • packaging: 25 g
  • storage: 4°C, stored under nitrogen; in solvent -80°C for 6 months or -20°C for 1 month

Catalog No. 50-002-26194 Supplier Medchemexpress LLC Supplier No. HY11668825G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Hydroxy-4-(methylthio)butyric acid (CAS 583-91-5) is the hydroxy analogue of methionine used as a biochemical reagent and reference standard for laboratory research. It is supplied as a light yellow to brown liquid and is intended for research applications under appropriate storage conditions.

  • hydroxy analogue of methionine (CAS 583-91-5)
  • molecular formula C5H10O3S and molecular weight 150.20 g/mol
  • appearance: light yellow to brown liquid
  • purity ≥88.0%
  • packaging: 25 g
  • storage: 4°C, stored under nitrogen; in solvent -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.