Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxy-4-(methylthio)butyric Acid (65-72% in Water), TCI America™
SDP

Catalog No. H065425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H065425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H065425G Supplier TCI America Supplier No. H065425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 583-91-5
Molecular Formula C10H18CaO6S2
Molecular Weight (g/mol) 338.45
MDL Number MFCD00070490
InChI Key ABRVDWASZFDIEH-UHFFFAOYNA-L
PubChem CID 11427
IUPAC Name calcium bis(2-hydroxy-4-(methylsulfanyl)butanoate)
SMILES [Ca++].CSCCC(O)C([O-])=O.CSCCC(O)C([O-])=O

Specifications

Quantity 25 g
Formula Weight 150.19
Physical Form Liquid
Chemical Name or Material 2-Hydroxy-4-(methylthio)butyric Acid (65-72% in Water)
RTECSNumber ET4731500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.