Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(Hydroxy(4-phenoxyphenyl)methylene)malononitrile | 330792-68-2 | 95.1% | C16H10N2O2 | 500 MG
SDP

Catalog No. 5000434411
Encompass_Preferred

2-(Hydroxy(4-phenoxyphenyl)methylene)malononitrile is a chemical compound primarily utilized as a drug intermediate in the synthesis of various active pharmaceutical compounds.

  • Molecular weight: 262.27
  • Appearance: Solid
  • Storage: Powder at -20°C for 3 years or 4°C for 2 years; in solvent at -80°C for 6 months or -20°C for 1 month.

Catalog No. 50-004-34411 Supplier Medchemexpress LLC Supplier No. HY78855500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-(Hydroxy(4-phenoxyphenyl)methylene)malononitrile is a chemical compound primarily utilized as a drug intermediate in the synthesis of various active pharmaceutical compounds.

  • Molecular weight: 262.27
  • Appearance: Solid
  • Storage: Powder at -20°C for 3 years or 4°C for 2 years; in solvent at -80°C for 6 months or -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.