Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(Hydroxy(4-phenoxyphenyl)methylene)malononitrile | 330792-68-2 | MFCD20270362 | >=98% | C16H10N2O2 | 5 G
SDP

Catalog No. 5000434894
Encompass_Preferred

2-(Hydroxy(4-phenoxyphenyl)methylene)malononitrile is a useful research chemical and drug intermediate for the synthesis of various active compounds, including pyrazolopyrimidines as protein kinase inhibitors. This compound appears as a solid and is stable under recommended storage conditions.

  • Appearance: Solid, light yellow to green yellow
  • Melting/freezing point: 160-161.2°C
  • Stability: Stable under recommended storage conditions
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • Safety: Harmful if swallowed; causes skin and serious eye irritation; may cause respiratory irritation

Catalog No. 50-004-34894 Supplier Medchemexpress LLC Supplier No. HY788555G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-(Hydroxy(4-phenoxyphenyl)methylene)malononitrile is a useful research chemical and drug intermediate for the synthesis of various active compounds, including pyrazolopyrimidines as protein kinase inhibitors. This compound appears as a solid and is stable under recommended storage conditions.

  • Appearance: Solid, light yellow to green yellow
  • Melting/freezing point: 160-161.2°C
  • Stability: Stable under recommended storage conditions
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • Safety: Harmful if swallowed; causes skin and serious eye irritation; may cause respiratory irritation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.