Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxy-5-iodopyridine, 97%, Thermo Scientific™

Catalog No. AC436370010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC436370010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC436370010 Supplier Thermo Scientific Chemicals Supplier No. 436370010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13472-79-2
Molecular Formula C5H4INO
Molecular Weight (g/mol) 220.99
InChI Key ZDJUNNCVIDKJAN-UHFFFAOYSA-N
Synonym 2-hydroxy-5-iodopyridine, 5-iodopyridin-2-ol, 5-iodopyridin-2 1h-one, 5-iodo-pyridin-2-ol, 5-iodo-2-pyridone, 2 1h-pyridinone, 5-iodo, 5-iodo-2-pyridinol, 5-jod-2-pyridon, 5-iodo-2-pyridol, 5-iodopyridin-2-one
PubChem CID 459500
IUPAC Name 5-iodo-1H-pyridin-2-one
SMILES C1=CC(=O)NC=C1I

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 186.7°C to 190.1°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 220.99
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-5-iodopyridine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.