Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2'-Hydroxy-5'-methylacetanilide 98.0+%, TCI America™
SDP

Catalog No. H14425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14425G 5 g
H144225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H14425G Supplier TCI America Supplier No. H14425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6375-17-3
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00020183
InChI Key DSEQJUPGRWESKP-UHFFFAOYSA-N
Synonym 2-acetamido-4-methylphenol, 2-hydroxy-5-methylacetanilide, n-2-hydroxy-5-methylphenyl acetamide, 2'-hydroxy-5'-methylacetanilide, acetamide, n-2-hydroxy-5-methylphenyl, 2-acetamido-4-cresol, n1-2-hydroxy-5-methylphenyl acetamide, 2-acetamido-p-cresol, 6'-hydroxy-m-acetotoluidide, n-2-hydroxy-5-methyl-phenyl acetamide
PubChem CID 292583
IUPAC Name N-(2-hydroxy-5-methylphenyl)acetamide
SMILES CC(=O)NC1=CC(C)=CC=C1O

Specifications

Melting Point 160°C
Color Yellow
Quantity 5 g
Formula Weight 165.19
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Hydroxy-5′-methylacetanilide
EINECSNumber (3)-0708
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.