Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2'-Hydroxy-5'-methylpropiophenone 98.0+%, TCI America™
SDP

Catalog No. H14941G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14941G 1 g
H14945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H14941G Supplier TCI America Supplier No. H14941G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 938-45-4
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD01098830
InChI Key CXZJBPYDVCLMFX-UHFFFAOYSA-N
Synonym 2-Propionyl-p-cresol
PubChem CID 70311
IUPAC Name 1-(2-hydroxy-5-methylphenyl)propan-1-one
SMILES CCC(=O)C1=CC(C)=CC=C1O

Specifications

Melting Point 2°C
Color Yellow
Quantity 1 g
Formula Weight 164.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2′-Hydroxy-5′-methylpropiophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.