Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2'-Hydroxy-5'-nitroacetophenone 98.0+%, TCI America™
SDP

Catalog No. H13785G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13785G 5 g
H137825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H13785G Supplier TCI America Supplier No. H13785G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1450-76-6
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.147
MDL Number MFCD00463816
InChI Key LNCBPUWMGYOISS-UHFFFAOYSA-N
PubChem CID 248079
SMILES CC(=O)C1=C(C=CC(=C1)[N+](=O)[O-])O

Specifications

Melting Point 102°C
Quantity 5 g
Formula Weight 181.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Hydroxy-5′-nitroacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.