Learn More
Medchemexpress LLC 2-Hydroxy-5-nitroben | 96-97-9 | 100g
SDP
Catalog No. 5000302331
Encompass_Preferred
2-Hydroxy-5-nitrobenzoic acid 99% is used as an HPLC analytical standard pH indicator and heavy metal detection agent and is also used in the study of phenolic compounds oxidation
Catalog No. 50-003-02331 Supplier Medchemexpress LLC Supplier No. HYW016352100G
2-Hydroxy-5-nitrobenzoic acid 99% is used as an HPLC analytical standard pH indicator and heavy metal detection agent and is also used in the study of phenolic compounds oxidation