Learn More
Medchemexpress LLC 2-Hydroxy-5-nitrobenzoic acid | 96-97-9 | MFCD00007338 | 96.8% | 183.12 g/mol | C7H5NO5 | 25 G
SDP
Catalog No. 5000304148
Encompass_Preferred
2-Hydroxy-5-nitrobenzoic acid is an analytical-grade nitrosalicylic acid commonly used in chromatography and chemical analysis. With an assay reported near 96.8%, it is provided with supporting documentation (COA, SDS, and chromatographic data) and is suitable for laboratory analytical workflows and research on phenolic oxidation.
Catalog No. 50-003-04148 Supplier Medchemexpress LLC Supplier No. 25GHYW016352
2-Hydroxy-5-nitrobenzoic acid is an analytical-grade nitrosalicylic acid commonly used in chromatography and chemical analysis. With an assay reported near 96.8%, it is provided with supporting documentation (COA, SDS, and chromatographic data) and is suitable for laboratory analytical workflows and research on phenolic oxidation.