Learn More
Medchemexpress LLC HY-W017100 100mg Medchemexpress, 2-Hydroxy-6-methoxybenzoic acid CAS:3147-64-6 Purity:>98%
SDP
Catalog No. 501934627
Encompass_Preferred
Medchemexpress, HY-W017100 100mg 2-Hydroxy-6-methoxybenzoic acid CAS:3147-64-6 2-Hydroxy-6-methoxybenzoic acid can be used for the determination of acetylsalicylic acid and its major metabolite, salicylic acid, in animal plasma. 2-Hydroxy-6-methoxybenzoic acid exhibits significant analgesic effects [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4627 Supplier Medchemexpress LLC Supplier No. HYW017100100MG
Medchemexpress, HY-W017100 100mg 2-Hydroxy-6-methoxybenzoic acid CAS:3147-64-6 2-Hydroxy-6-methoxybenzoic acid can be used for the determination of acetylsalicylic acid and its major metabolite, salicylic acid, in animal plasma. 2-Hydroxy-6-methoxybenzoic acid exhibits significant analgesic effects [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.