Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-hydroxy-6-methoxybenzoic acid | 3147-64-6 | MFCD00674090 | 99.3% | 168.15 g/mol | C8H8O4 | 1 G
SDP

Catalog No. 5000299860
Encompass_Preferred

2-hydroxy-6-methoxybenzoic acid is a benzoic acid derivative with reported analgesic activity used in biochemical and pharmacology research. Supplied as a solid research chemical, it is suitable for preclinical studies and small-molecule screening.

  • High purity (99.34%).
  • Molecular weight 168.15 g/mol.
  • Chemical formula C8H8O4.
  • White to off-white solid appearance.
  • Soluble in DMSO (~100 mg/mL) with ultrasonic aid.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years).
  • Available in multiple pack sizes including 1 g.

Catalog No. 50-002-99860 Supplier Medchemexpress LLC Supplier No. HYW0171001G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

2-hydroxy-6-methoxybenzoic acid is a benzoic acid derivative with reported analgesic activity used in biochemical and pharmacology research. Supplied as a solid research chemical, it is suitable for preclinical studies and small-molecule screening.

  • High purity (99.34%).
  • Molecular weight 168.15 g/mol.
  • Chemical formula C8H8O4.
  • White to off-white solid appearance.
  • Soluble in DMSO (~100 mg/mL) with ultrasonic aid.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years).
  • Available in multiple pack sizes including 1 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.