Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-hydroxy-6-methoxybenzoic acid | 3147-64-6 | MFCD00674090 | 99.3% | 168.15 g/mol | C8H8O4 | 100 G
SDP

Catalog No. 5000299513
Encompass_Preferred

2-Hydroxy-6-methoxybenzoic acid (6-methoxysalicylic acid) is a small aromatic carboxylic acid used as a research reagent and synthetic intermediate. Supplied as a white to off-white solid with a molecular weight of 168.15 g/mol, it is used in biochemical and medicinal chemistry applications and is provided with solubility and storage guidance for laboratory use.

  • High purity (reported 99.34%) for reliable analytical and synthetic work.
  • White to off-white solid form, easy to weigh and handle.
  • Soluble in DMSO (100 mg/mL) and formulatable for in vivo studies.
  • Stable under recommended storage conditions for extended shelf life.
  • Available in multiple pack sizes and ready-to-use DMSO solutions.
  • SMILES and structural identifiers provided for cheminformatics use.

Catalog No. 50-002-99513 Supplier Medchemexpress LLC Supplier No. HYW017100100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

2-Hydroxy-6-methoxybenzoic acid (6-methoxysalicylic acid) is a small aromatic carboxylic acid used as a research reagent and synthetic intermediate. Supplied as a white to off-white solid with a molecular weight of 168.15 g/mol, it is used in biochemical and medicinal chemistry applications and is provided with solubility and storage guidance for laboratory use.

  • High purity (reported 99.34%) for reliable analytical and synthetic work.
  • White to off-white solid form, easy to weigh and handle.
  • Soluble in DMSO (100 mg/mL) and formulatable for in vivo studies.
  • Stable under recommended storage conditions for extended shelf life.
  • Available in multiple pack sizes and ready-to-use DMSO solutions.
  • SMILES and structural identifiers provided for cheminformatics use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.