Learn More
Medchemexpress LLC 2-hydroxy-6-methoxybenzoic acid | 3147-64-6 | MFCD00674090 | 99.3% | 168.15 g/mol | C8H8O4 | 5 G
SDP
Catalog No. 5000300147
Encompass_Preferred
2-Hydroxy-6-methoxybenzoic acid is a salicylic-acid derivative used as a research reagent in chemical, analytical, and biological studies. It has molecular formula C8H8O4 and a molecular weight of 168.15 g/mol. The compound is supplied with high reported purity and is commonly used as an analytical standard, synthetic intermediate, and reference material in laboratory workflows.
  • High purity (99.3%).
  • Suitable as an analytical standard and reference material.
  • Useful as a synthetic intermediate in organic synthesis.
  • Available in solid and solution formats for convenient use.
  • Molecular formula and weight provided for identification and calculations.
Catalog No. 50-003-00147 Supplier Medchemexpress LLC Supplier No. HYW0171005G
2-Hydroxy-6-methoxybenzoic acid is a salicylic-acid derivative used as a research reagent in chemical, analytical, and biological studies. It has molecular formula C8H8O4 and a molecular weight of 168.15 g/mol. The compound is supplied with high reported purity and is commonly used as an analytical standard, synthetic intermediate, and reference material in laboratory workflows.
  • High purity (99.3%).
  • Suitable as an analytical standard and reference material.
  • Useful as a synthetic intermediate in organic synthesis.
  • Available in solid and solution formats for convenient use.
  • Molecular formula and weight provided for identification and calculations.