Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxy-6-methyl-3-nitropyridine 98.0+%, TCI America™
SDP

Catalog No. H14981G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14981G 1 g
H14985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H14981G Supplier TCI America Supplier No. H14981G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 39745-39-6
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.125
MDL Number MFCD03001414
InChI Key YVYDGIGILRUPED-UHFFFAOYSA-N
Synonym 6-methyl-3-nitropyridin-2-ol, 2-hydroxy-6-methyl-3-nitropyridine, 6-hydroxy-5-nitro-2-picoline, 2-hydroxy-3-nitro-6-methylpyridine, 2-hydroxy-3-nitro-6-picoline, 6-methyl-3-nitro-2 1h-pyridinone, 6-methyl-3-nitro-2-pyridone, 3-nitro-6-methylpyridin-2-ol, 2-hydroxy-3-nitro-methylpyridine, 2 1h-pyridinone, 6-methyl-3-nitro
PubChem CID 543029
IUPAC Name 6-methyl-3-nitro-1H-pyridin-2-one
SMILES CC1=CC=C(C(=O)N1)[N+](=O)[O-]

Specifications

Melting Point 217°C
Quantity 1 g
Formula Weight 154.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-6-methyl-3-nitropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.