Learn More
Cayman Chemical 2-hydroxy FlutamIde 50mg
SDP
Catalog No. 502972823
Encompass_Preferred
The major metabolite formed during the metabolism of the non-steroidal antiandrogen flutamide by cytochrome P450 isoforms CYP1A2 and CYP3A4; inhibits the binding of testosterone to the nuclear androgen receptor (IC50 = ~300-900), blocking the expression of AR target genes
Catalog No. 50-297-2823 Supplier Cayman Chemical Supplier No. 1527150 MG
The major metabolite formed during the metabolism of the non-steroidal antiandrogen flutamide by cytochrome P450 isoforms CYP1A2 and CYP3A4; inhibits the binding of testosterone to the nuclear androgen receptor (IC50 = ~300-900), blocking the expression of AR target genes