Learn More
Cayman Chemical (±)-2-hydroxy Ibuprofen, Purity: ≥95%, CAS: 51146-55-5, 1 mg
SDP
Catalog No. 502933902
Encompass_Preferred
A metabolite of ibuprofen
Catalog No. 50-293-3902 Supplier Cayman Chemical Supplier No. 265951 MG
A metabolite of ibuprofen