Learn More
Cayman Chemical 2-hydroxy Lauric Acid, Purity: ≥98%, CAS: 2984-55-6, 50 mg
SDP
Catalog No. 502936240
Encompass_Preferred
A hydroxy fatty acid; a partial agonist of FFAR1/GPR40 and GPR85 receptors in vitro (EC50s = 6.7 and 12.8 μM, respectively); inhibits bovine hepatic ligase (Ki = 4.4 μM); inhibits medium-chain acyl-CoA synthetase by 48% in mouse kidney mitochondria (40 μM)
Catalog No. 50-293-6240 Supplier Cayman Chemical Supplier No. 2458650 MG
A hydroxy fatty acid; a partial agonist of FFAR1/GPR40 and GPR85 receptors in vitro (EC50s = 6.7 and 12.8 μM, respectively); inhibits bovine hepatic ligase (Ki = 4.4 μM); inhibits medium-chain acyl-CoA synthetase by 48% in mouse kidney mitochondria (40 μM)