Learn More
Cayman Chemical 2-hydroxy Myristic Acid, Purity: ≥98%, CAS: 2507-55-3, 50 mg
SDP
Catalog No. 502896069
Encompass_Preferred
A hydroxy fatty acid; inhibits cleavage between the enterovirus capsid proteins VP4 and VP2; inhibits Junin and Tacaribe viral replication (IC50s = 20.1 and 14.2 μM, respectively); previously characterized as a weak inhibitor of peptide myristoylation (Ki = 200 μM) but has been shown to be inactive in ARL1 cells when used at 100 μM
Catalog No. 50-289-6069 Supplier Cayman Chemical Supplier No. 9039050 MG
A hydroxy fatty acid; inhibits cleavage between the enterovirus capsid proteins VP4 and VP2; inhibits Junin and Tacaribe viral replication (IC50s = 20.1 and 14.2 μM, respectively); previously characterized as a weak inhibitor of peptide myristoylation (Ki = 200 μM) but has been shown to be inactive in ARL1 cells when used at 100 μM