Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-hydroxy-N,N-dimethyl-3-[[2-[[(1R)-1-[4-(1-methylethyl)-2-furanyl]propyl]amino]...benzamide | 473728-58-4 | >98.0% | 425.5 g/mol | C23H27N3O5 | 1 ML
SDP

Catalog No. 5000215167
Encompass_Preferred

(Rac)-SCH 563705 is a racemic small-molecule antagonist of CXCR2 and CXCR1 supplied as a 10 mM solution in a 1 mL vial for research use. It has molecular formula C23H27N3O5, a molecular weight of 425.5 g/mol, and CAS number 473728-58-4. The compound is used to study chemokine receptor signaling and inflammatory responses in biological assays.

  • Racemic CXCR2/CXCR1 antagonist for chemokine receptor research.
  • Supplied as a 10 mM solution in a 1 mL vial for convenient dosing.
  • High purity (>98.0%) suitable for biological assays.
  • Molecular formula C23H27N3O5 and molecular weight 425.5 g/mol.
  • CAS number 473728-58-4 for unambiguous identification.

Catalog No. 50-002-15167 Supplier Medchemexpress LLC Supplier No. HY10011B10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Rac)-SCH 563705 is a racemic small-molecule antagonist of CXCR2 and CXCR1 supplied as a 10 mM solution in a 1 mL vial for research use. It has molecular formula C23H27N3O5, a molecular weight of 425.5 g/mol, and CAS number 473728-58-4. The compound is used to study chemokine receptor signaling and inflammatory responses in biological assays.

  • Racemic CXCR2/CXCR1 antagonist for chemokine receptor research.
  • Supplied as a 10 mM solution in a 1 mL vial for convenient dosing.
  • High purity (>98.0%) suitable for biological assays.
  • Molecular formula C23H27N3O5 and molecular weight 425.5 g/mol.
  • CAS number 473728-58-4 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.