Learn More
Cayman Chemical 2-hydroxy Palmitic Acid, Purity: ≥95%, CAS: 764-67-0, 50 mg
SDP
Catalog No. 502994720
Encompass_Preferred
An intermediate in the metabolism of phytosphingosine; found in yeast fermentations
Catalog No. 50-299-4720 Supplier Cayman Chemical Supplier No. 2267950 MG
An intermediate in the metabolism of phytosphingosine; found in yeast fermentations