Learn More
Cayman Chemical 2-hydroxy Stearic Acid, Purity: ≥98%, CAS: 629-22-1, 50 mg
SDP
Catalog No. 502933417
Encompass_Preferred
A hydroxy fatty acid; has been found in humans and U. armoricana; reduces growth of EAT cells (100 μM); elevated in human joint synovial fluid after ankle fracture and in patients with Buruli ulcer disease
Catalog No. 50-293-3417 Supplier Cayman Chemical Supplier No. 2459050 MG
A hydroxy fatty acid; has been found in humans and U. armoricana; reduces growth of EAT cells (100 μM); elevated in human joint synovial fluid after ankle fracture and in patients with Buruli ulcer disease