Learn More
Cayman Chemical 2'-Hydroxyacetophenone, Purity: ≥98%, CAS: 118-93-4, 1 kg
SDP
Catalog No. 502915835
Encompass_Preferred
An acetophenone synthetic building block; has been used in the synthesis of hydroxychalcone derivatives with inhibitory activity towards COX-2
Catalog No. 50-291-5835 Supplier Cayman Chemical Supplier No. 374181 KG
An acetophenone synthetic building block; has been used in the synthesis of hydroxychalcone derivatives with inhibitory activity towards COX-2