Learn More
Cayman Chemical 2-Hydroxyadipic Acid, Purity: ≥95%, CAS: 18294-85-4, 10 mg
SDP
Catalog No. 502912311
Encompass_Preferred
A dicarboxylic fatty acid; levels are decreased in tumor tissue from patients with esophageal squamous cell carcinoma; levels are increased in patients with 2-ketoadipic aciduria
Catalog No. 50-291-2311 Supplier Cayman Chemical Supplier No. 3648210 MG
A dicarboxylic fatty acid; levels are decreased in tumor tissue from patients with esophageal squamous cell carcinoma; levels are increased in patients with 2-ketoadipic aciduria