Learn More
Medchemexpress LLC 2-Hydroxybenzoic acid (Standard) | 69-72-7 | 98.0% | 100 MG
SDP
Catalog No. 5000407168
Encompass_Preferred
Salicylic acid (Standard) is the analytical standard of Salicylic acid, intended for research and analytical applications. It inhibits cyclo-oxygenase-2 (COX-2) activity independently of transcription factor (NF-κB) activation.
  • Intended for research and analytical applications
  • Inhibits cyclo-oxygenase-2 (COX-2) activity
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS
Catalog No. 50-004-07168 Supplier Medchemexpress LLC Supplier No. HYB0167R100MG
Salicylic acid (Standard) is the analytical standard of Salicylic acid, intended for research and analytical applications. It inhibits cyclo-oxygenase-2 (COX-2) activity independently of transcription factor (NF-κB) activation.
  • Intended for research and analytical applications
  • Inhibits cyclo-oxygenase-2 (COX-2) activity
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS