Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Hydroxybenzonitrile | 611-20-1 | 99.99% | 119.12 | 250 G
SDP

Catalog No. 5000366377
Encompass_Preferred

2-Hydroxybenzonitrile is a biochemical reagent designed for life science research. It serves as a versatile biological material or organic compound for various laboratory applications.

  • Used for life science related research
  • Solid appearance
  • Off-white to light yellow color
  • Stable under recommended storage conditions
  • Melting point of 92-95°C
  • Boiling point of 149°C at 14 mmHg

Catalog No. 50-003-66377 Supplier Medchemexpress LLC Supplier No. 250GHYW012869
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Hydroxybenzonitrile is a biochemical reagent designed for life science research. It serves as a versatile biological material or organic compound for various laboratory applications.

  • Used for life science related research
  • Solid appearance
  • Off-white to light yellow color
  • Stable under recommended storage conditions
  • Melting point of 92-95°C
  • Boiling point of 149°C at 14 mmHg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.