Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxybenzophenone 99%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
25g
Packaging:
Glass bottle
117-99-7
C13H10O2
198.22
MFCD00002216
HJIAMFHSAAEUKR-UHFFFAOYSA-N
2-hydroxybenzophenone,o-benzoylphenol,2-hydroxyphenyl phenyl methanone,o-hydroxybenzophenone,benzophenone, 2-hydroxy,methanone, 2-hydroxyphenyl phenyl,phenyl 2-hydroxyphenyl ketone,unii-lqv22w4v05,ortho-hydroxybenzophenone,2-hydroxyphenyl phenylmethanone
8348
(2-hydroxyphenyl)-phenylmethanone
C1=CC=C(C=C1)C(=O)C2=CC=CC=C2O
117-99-7
C13H10O2
198.22
MFCD00002216
HJIAMFHSAAEUKR-UHFFFAOYSA-N
2-hydroxybenzophenone,o-benzoylphenol,2-hydroxyphenyl phenyl methanone,o-hydroxybenzophenone,benzophenone, 2-hydroxy,methanone, 2-hydroxyphenyl phenyl,phenyl 2-hydroxyphenyl ketone,unii-lqv22w4v05,ortho-hydroxybenzophenone,2-hydroxyphenyl phenylmethanone
8348
(2-hydroxyphenyl)-phenylmethanone
C1=CC=C(C=C1)C(=O)C2=CC=CC=C2O
Boiling Point 171°C to 173°C (12.0mmHg)
Flash Point >112°C
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 25g
Melting Point 37°C to 40°C
Color Yellow
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula HOC6H4COC6H5
Beilstein 08, 155
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol
Formula Weight 198.22
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material 2-Hydroxybenzophenone, 99%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 204-226-2

TSCA : TSCA