Learn More
Selleck Chemical LLC 2-Hydroxybenzylamine
SDP
Catalog No. 503136667
Encompass_Preferred
2-Hydroxybenzylamine (2-(Aminomethyl)phenol 2-HOBA NSC 127870) is a naturally occurring compound found in buckwheat and is a selective scavenger of dicarbonyl electrophiles that protects proteins and lipids from being modified by these electrophiles 2-HOBA reduces inflammation and plaque apoptotic cells and promotes efferocytosis and features of stable plaques 2-HOBA has potential for use as a nutrition supplement due to its ability to protect against the damaging effects of oxidative stress
Catalog No. 50-313-6667 Supplier Selleck Chemical LLC Supplier No. E002510MM/1ML
2-Hydroxybenzylamine (2-(Aminomethyl)phenol 2-HOBA NSC 127870) is a naturally occurring compound found in buckwheat and is a selective scavenger of dicarbonyl electrophiles that protects proteins and lipids from being modified by these electrophiles 2-HOBA reduces inflammation and plaque apoptotic cells and promotes efferocytosis and features of stable plaques 2-HOBA has potential for use as a nutrition supplement due to its ability to protect against the damaging effects of oxidative stress