Learn More
Selleck Chemical LLC 2-Hydroxybutyric acid
Supplier: Selleck Chemical LLC S611925MG
2-Hydroxybutyric acid ( -hydroxybutyrate) is converted from 2-Aminobutyric acid It is produced in mammalian tissues (principally hepatic) that catabolize L-threonine or synthesize glutathione