Learn More
Selleck Chemical LLC 2-Hydroxybutyric acid
SDP
Catalog No. 5000004400
Encompass_Preferred
2-Hydroxybutyric acid ( -hydroxybutyrate) is converted from 2-Aminobutyric acid It is produced in mammalian tissues (principally hepatic) that catabolize L-threonine or synthesize glutathione
Catalog No. 50-000-04400 Supplier Selleck Chemical LLC Supplier No. S611950MG
2-Hydroxybutyric acid ( -hydroxybutyrate) is converted from 2-Aminobutyric acid It is produced in mammalian tissues (principally hepatic) that catabolize L-threonine or synthesize glutathione