Learn More
TARGETMOL CHEMICALS INC 2-Hydroxybutyric acid 500MG
SDP
Catalog No. 502878845
Encompass_Preferred
Also available in 1 mL, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. 2-Hydroxybutyric acid (alpha-Hydroxybutyric acid) is a product of protein metabolism and serves as a biomarker for type 2 diabetes and pre-eclampsia. Purity 98%
Catalog No. 50-287-8845 Supplier TARGETMOL CHEMICALS INC Supplier No. T4322500MG
Also available in 1 mL, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. 2-Hydroxybutyric acid (alpha-Hydroxybutyric acid) is a product of protein metabolism and serves as a biomarker for type 2 diabetes and pre-eclampsia. Purity 98%