Learn More
Medchemexpress LLC 2-Hydroxybutyric acid | 600-15-7 | 98.0% | 104.10 | 100 MG
SDP
Catalog No. 5000436642
Encompass_Preferred
2-Hydroxybutyric acid is an endogenous metabolite derived from 2-Aminobutyric acid. It is identified as a potential biomarker for type 2 diabetes and preeclampsia. This compound has demonstrated the ability to prevent acetaminophen (AP)-induced liver injury in studies, enhancing cell viability and reducing AP-induced cytotoxicities in various cell lines and in vivo models. It also plays a role as a coproduct of insulin resistance.
  • Potential biomarker for type 2 diabetes and preeclampsia.
  • Prevents acetaminophen-induced liver injury.
  • Enhances cell viabilities and reduces AP-induced cytotoxicities.
  • Normalizes gene expressions related to inflammation, oxidative stress, cell proliferation, and DNA damage.
  • Functions as a coproduct of insulin resistance.
Catalog No. 50-004-36642 Supplier Medchemexpress LLC Supplier No. HY113381100MG
2-Hydroxybutyric acid is an endogenous metabolite derived from 2-Aminobutyric acid. It is identified as a potential biomarker for type 2 diabetes and preeclampsia. This compound has demonstrated the ability to prevent acetaminophen (AP)-induced liver injury in studies, enhancing cell viability and reducing AP-induced cytotoxicities in various cell lines and in vivo models. It also plays a role as a coproduct of insulin resistance.
  • Potential biomarker for type 2 diabetes and preeclampsia.
  • Prevents acetaminophen-induced liver injury.
  • Enhances cell viabilities and reduces AP-induced cytotoxicities.
  • Normalizes gene expressions related to inflammation, oxidative stress, cell proliferation, and DNA damage.
  • Functions as a coproduct of insulin resistance.