Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-hydroxychalcone | 644-78-0 | 98.1% | 1 G
SDP

Catalog No. 5000233025
Encompass_Preferred

2-hydroxychalcone is a small-molecule chalcone used as a research reagent; it has reported antioxidant activity and modulates NF-κB and Bcl-2 pathways. The compound is supplied as a light yellow solid with high purity suitable for in vitro biochemical studies.

  • Purity: 98.08% (reported).
  • Molecular weight: 224.25 g/mol; formula C15H12O2.
  • Reported activities: antioxidant, NF-κB inhibition, Bcl-2 downregulation.
  • Solubility: DMSO or ethanol up to 100 mg/mL (may require ultrasonication).
  • Storage: store at -20°C protected from light; in solvent store at -80°C for long-term stability.
  • Available formats: 10 mM solutions in DMSO and solids in 500 mg, 1 g, 5 g, and 10 g quantities.

Catalog No. 50-002-33025 Supplier Medchemexpress LLC Supplier No. HY1199311G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-hydroxychalcone is a small-molecule chalcone used as a research reagent; it has reported antioxidant activity and modulates NF-κB and Bcl-2 pathways. The compound is supplied as a light yellow solid with high purity suitable for in vitro biochemical studies.

  • Purity: 98.08% (reported).
  • Molecular weight: 224.25 g/mol; formula C15H12O2.
  • Reported activities: antioxidant, NF-κB inhibition, Bcl-2 downregulation.
  • Solubility: DMSO or ethanol up to 100 mg/mL (may require ultrasonication).
  • Storage: store at -20°C protected from light; in solvent store at -80°C for long-term stability.
  • Available formats: 10 mM solutions in DMSO and solids in 500 mg, 1 g, 5 g, and 10 g quantities.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.