Learn More
Medchemexpress LLC 2-Hydroxycinnamic acid | 614-60-8 | 98.71% | 164.16 | 25 MG
SDP
Catalog No. 5000452040
Encompass_Preferred
2-Hydroxycinnamic acid (Standard) is the analytical standard of 2-Hydroxycinnamic acid, intended for research and analytical applications. It is a phenolic acid with antimicrobial and antioxidant properties.
  • Antimicrobial activity against Staphylococcus aureus, not susceptible to drug resistance.
  • Shows inhibitory effects on infection of HIV/SARS-CoV S pseudovirus with an IC50 of 0.3 mM.
  • Has neuroprotective and antitumor activity.
  • The compound is an analytical standard, commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-004-52040 Supplier Medchemexpress LLC Supplier No. 25MGHYW012531R
2-Hydroxycinnamic acid (Standard) is the analytical standard of 2-Hydroxycinnamic acid, intended for research and analytical applications. It is a phenolic acid with antimicrobial and antioxidant properties.
  • Antimicrobial activity against Staphylococcus aureus, not susceptible to drug resistance.
  • Shows inhibitory effects on infection of HIV/SARS-CoV S pseudovirus with an IC50 of 0.3 mM.
  • Has neuroprotective and antitumor activity.
  • The compound is an analytical standard, commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.