Learn More
Medchemexpress LLC 2-Hydroxycinnamic acid | 614-60-8 | 99.9% | 164.16 | 1 G
SDP
Catalog No. 5000452933
Encompass_Preferred
2-Hydroxycinnamic acid is a phenolic acid possessing antimicrobial and antioxidant properties. It exhibits antimicrobial activity against Staphylococcus aureus without succumbing to drug resistance. Furthermore, it demonstrates inhibitory effects on HIV/SARS-CoV S pseudovirus infection with an IC50 of 0.3 mM. The compound also presents neuroprotective and antitumor activities.
  • Antimicrobial properties: Effective against Staphylococcus aureus, including drug-resistant strains.
  • Antioxidant properties: Acts as an antioxidant.
  • Antiviral activity: Inhibits infection of HIV/SARS-CoV S pseudovirus.
  • Neuroprotective activity: Offers protective effects on neurons.
  • Antitumor activity: Shows potential in combating tumors.
Catalog No. 50-004-52933 Supplier Medchemexpress LLC Supplier No. HYW0125311G
2-Hydroxycinnamic acid is a phenolic acid possessing antimicrobial and antioxidant properties. It exhibits antimicrobial activity against Staphylococcus aureus without succumbing to drug resistance. Furthermore, it demonstrates inhibitory effects on HIV/SARS-CoV S pseudovirus infection with an IC50 of 0.3 mM. The compound also presents neuroprotective and antitumor activities.
  • Antimicrobial properties: Effective against Staphylococcus aureus, including drug-resistant strains.
  • Antioxidant properties: Acts as an antioxidant.
  • Antiviral activity: Inhibits infection of HIV/SARS-CoV S pseudovirus.
  • Neuroprotective activity: Offers protective effects on neurons.
  • Antitumor activity: Shows potential in combating tumors.