Learn More
Medchemexpress LLC 2-Hydroxycinnamic acid | 614-60-8 | HY-W012531 | 99.9% | 164.16 | 25 G
SDP
Catalog No. 5000451220
Encompass_Preferred
2-Hydroxycinnamic acid is a phenolic acid with antimicrobial and antioxidant properties. It demonstrates antimicrobial activity against *Staphylococcus aureus* without developing resistance, and shows inhibitory effects on HIV/SARS-CoV S pseudovirus infection. Additionally, it exhibits neuroprotective and antitumor activity.
  • Possesses antimicrobial and antioxidant properties.
  • Exhibits antimicrobial activity against *Staphylococcus aureus*.
  • Shows inhibitory effects on HIV/SARS-CoV S pseudovirus infection.
  • Has neuroprotective activity.
  • Demonstrates antitumor activity.
Catalog No. 50-004-51220 Supplier Medchemexpress LLC Supplier No. 25GHYW012531
2-Hydroxycinnamic acid is a phenolic acid with antimicrobial and antioxidant properties. It demonstrates antimicrobial activity against *Staphylococcus aureus* without developing resistance, and shows inhibitory effects on HIV/SARS-CoV S pseudovirus infection. Additionally, it exhibits neuroprotective and antitumor activity.
  • Possesses antimicrobial and antioxidant properties.
  • Exhibits antimicrobial activity against *Staphylococcus aureus*.
  • Shows inhibitory effects on HIV/SARS-CoV S pseudovirus infection.
  • Has neuroprotective activity.
  • Demonstrates antitumor activity.